Anti-UBE2E3 Rabbit Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: UBE2E3
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77721-573
- 77721-572
- Antigen symbol:
- UBE2E3
- UBE2E3
- Antigen synonyms:
- Ubiquitin-protein ligase E3|Ubiquitin-conjugating enzyme E2 E3|Ubiquitin conjugating enzyme E2E 3 (UBC4/5 homolog, yeast|UbcH9|Ubiquitin-conjugating enzyme E2-23 kDa|Ubiquitin conjugating enzyme E2 E3|Ubiquitin conjugating enzyme E2E 3|Ubc M2|Ubiquitin conjugating enzyme E2 23 kDa|UBC4/5 homolog|UBCE4|Ubiquitin carrier protein|UBE2 E3|Ubiquitin conjugating enzyme E2E 3 (homologous to yeast UBC4/5)|Homologous to yeast UBC4/5|UbcM2|UB2E3_HUMAN|UBE2E3|Ubiquitin protein ligase|Ubiquitin carrier protein E3|UBC E4|UBCH 9|UBCE 4|Ubiquitin conjugating enzyme UBCH9|Ubiquitin protein ligase E3|Ubiquitin conjugating enzyme UBCH 9
- Ubiquitin-protein ligase E3|Ubiquitin-conjugating enzyme E2 E3|Ubiquitin conjugating enzyme E2E 3 (UBC4/5 homolog, yeast|UbcH9|Ubiquitin-conjugating enzyme E2-23 kDa|Ubiquitin conjugating enzyme E2 E3|Ubiquitin conjugating enzyme E2E 3|Ubc M2|Ubiquitin conjugating enzyme E2 23 kDa|UBC4/5 homolog|UBCE4|Ubiquitin carrier protein|UBE2 E3|Ubiquitin conjugating enzyme E2E 3 (homologous to yeast UBC4/5)|Homologous to yeast UBC4/5|UbcM2|UB2E3_HUMAN|UBE2E3|Ubiquitin protein ligase|Ubiquitin carrier protein E3|UBC E4|UBCH 9|UBCE 4|Ubiquitin conjugating enzyme UBCH9|Ubiquitin protein ligase E3|Ubiquitin conjugating enzyme UBCH 9
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q969T4
- Q969T4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Ubiquitin-conjugating enzyme E2 E3 protein (2-128AA)
- Recombinant Human Ubiquitin-conjugating enzyme E2 E3 protein (2-128AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together