Anti-SSH1 Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SSH1
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77721-430
- 77721-429
- Antigen symbol:
- SSH1
- SSH1
- Antigen synonyms:
- SSH1_HUMAN|actin binding protein 1C|Gm1395|coronin|Coro1c|SSH-1L|slingshot like 1|mSSH 1L|Ssh1|SSH-like protein 1|hSSH-1L|AW551225|Protein phosphatase Slingshot homolog 1|slingshot homolog 1|Slingshot1|slingshot 1L|SSH 1L|Gm1394|SSH 1|OTTMUSP00000031415|Slingshot 1
- SSH1_HUMAN|actin binding protein 1C|Gm1395|coronin|Coro1c|SSH-1L|slingshot like 1|mSSH 1L|Ssh1|SSH-like protein 1|hSSH-1L|AW551225|Protein phosphatase Slingshot homolog 1|slingshot homolog 1|Slingshot1|slingshot 1L|SSH 1L|Gm1394|SSH 1|OTTMUSP00000031415|Slingshot 1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8WYL5
- Q8WYL5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Protein phosphatase Slingshot homolog 1 protein (928-1049AA)
- Recombinant Human Protein phosphatase Slingshot homolog 1 protein (928-1049AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together