Anti-TNFAIP8L1 Rabbit Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TNFAIP8L1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77721-359
- 77721-358
- Antigen symbol:
- TNFAIP8L1
- TNFAIP8L1
- Antigen synonyms:
- Tumor necrosis factor alpha-induced protein 8-like protein 1|TIPE1|TNFAIP8-like protein 1|Oxy-beta|TNFAIP8L1|TNF alpha-induced protein 8-like protein 1|Oxidative stress-regulated gene-beta
- Tumor necrosis factor alpha-induced protein 8-like protein 1|TIPE1|TNFAIP8-like protein 1|Oxy-beta|TNFAIP8L1|TNF alpha-induced protein 8-like protein 1|Oxidative stress-regulated gene-beta
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8WVP5
- Q8WVP5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Tumor necrosis factor alpha-induced protein 8-like protein 1 protein (1-186AA)
- Recombinant Human Tumor necrosis factor alpha-induced protein 8-like protein 1 protein (1-186AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together