Anti-RNF139 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20-1:200
Type:
Antigen: RNF139
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77721-314
- 77721-313
- Antigen symbol:
- RNF139
- RNF139
- Antigen synonyms:
- TRC8|RNF139|EC 2.3.2.27|E3 ubiquitin-protein ligase RNF139|RING finger protein 139|RING-type E3 ubiquitin transferase RNF139|Translocation in renal carcinoma on chromosome 8 protein
- TRC8|RNF139|EC 2.3.2.27|E3 ubiquitin-protein ligase RNF139|RING finger protein 139|RING-type E3 ubiquitin transferase RNF139|Translocation in renal carcinoma on chromosome 8 protein
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8WU17
- Q8WU17
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human E3 ubiquitin-protein ligase RNF139 protein (510-664AA)
- Recombinant Human E3 ubiquitin-protein ligase RNF139 protein (510-664AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together