Anti-ZDHHC1 Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ZDHHC1
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77721-309
- 77721-310
- Antigen symbol:
- ZDHHC1
- ZDHHC1
- Antigen synonyms:
- Probable palmitoyltransferase ZDHHC1|C16orf1|Zdhhc1|Zinc finger DHHC type containing 1|Zinc finger DHHC domain containing 1|ZNF377|HSU90653|Zinc finger DHHC domain-containing protein 1|DHHC domain-containing cysteine-rich protein 1|DHHC domain containing cysteine rich protein|DHHC-1|Zinc finger protein 377|ZDHC1_HUMAN
- Probable palmitoyltransferase ZDHHC1|C16orf1|Zdhhc1|Zinc finger DHHC type containing 1|Zinc finger DHHC domain containing 1|ZNF377|HSU90653|Zinc finger DHHC domain-containing protein 1|DHHC domain-containing cysteine-rich protein 1|DHHC domain containing cysteine rich protein|DHHC-1|Zinc finger protein 377|ZDHC1_HUMAN
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8WTX9
- Q8WTX9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Probable palmitoyltransferase ZDHHC1 protein (283-389AA)
- Recombinant Human Probable palmitoyltransferase ZDHHC1 protein (283-389AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together