Anti-SSH3 Rabbit Polyclonal Antibody (HRP)
New Product
Type:
Antigen: SSH3
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77721-270
- 77721-269
- Antigen symbol:
- SSH3
- SSH3
- Antigen synonyms:
- SSH3_HUMAN|Protein phosphatase Slingshot homolog 3|FLJ20515|Slingshot homolog 3|SSH-like protein 3|hSSH 3L|Slingshot 3|Slingshot3|SSH-3L|SSH3L|SSH 3|FLJ10928|hSSH-3L|hSSH3L|SSH 3L|Ssh3
- SSH3_HUMAN|Protein phosphatase Slingshot homolog 3|FLJ20515|Slingshot homolog 3|SSH-like protein 3|hSSH 3L|Slingshot 3|Slingshot3|SSH-3L|SSH3L|SSH 3|FLJ10928|hSSH-3L|hSSH3L|SSH 3L|Ssh3
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8TE77
- Q8TE77
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Protein phosphatase Slingshot homolog 3 protein (480-659AA)
- Recombinant Human Protein phosphatase Slingshot homolog 3 protein (480-659AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together