Anti-HORMAD2 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
About this item
Recommended dilution: IHC:1:20-1:200
Type:
Antigen: HORMAD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
2 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Recommended dilution: IHC:1:20-1:200
Type:
Antigen: HORMAD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications for Product Family
Specifications
- Catalog No:
- 77721-028
- 77721-027
- Antigen symbol:
- HORMAD2
- HORMAD2
- Antigen synonyms:
- RP23-329P17.4|MGC26710|HORM2_HUMAN|HORMA domain-containing protein 2|HORMA domain containing 2|CT46.2|4930529M09Rik|Hormad2
- RP23-329P17.4|MGC26710|HORM2_HUMAN|HORMA domain-containing protein 2|HORMA domain containing 2|CT46.2|4930529M09Rik|Hormad2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8N7B1
- Q8N7B1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human HORMA domain-containing protein 2 protein (14-307AA)
- Recombinant Human HORMA domain-containing protein 2 protein (14-307AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Product Family Options
Product Information
- SizePackagingAvailabilityPrice
- Supplier Number: A62857-50UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.77721-028Antigen symbolHORMAD2Antigen synonymsRP23-329P17.4|MGC26710|HORM2_HUMAN|HORMA domain-containing protein 2|HORMA domain containing 2|CT46.2|4930529M09Rik|Hormad2ClonalityPolyclonalConjugationUnconjugatedReactivityHumanHostRabbitGene IDQ8N7B1IsotypeIgGELISAYesImmunoChemistryYesImmunogenRecombinant Human HORMA domain-containing protein 2 protein (14-307AA)FormLiquidPurification>95%, Protein G purifiedStorage buffer0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4Storage temperatureUpon receipt, store at –20 or –80 °C. Avoid repeated freeze.Shipping temperatureShipped on blue ice - Supplier Number: A62857-100UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.77721-027Antigen symbolHORMAD2Antigen synonymsRP23-329P17.4|MGC26710|HORM2_HUMAN|HORMA domain-containing protein 2|HORMA domain containing 2|CT46.2|4930529M09Rik|Hormad2ClonalityPolyclonalConjugationUnconjugatedReactivityHumanHostRabbitGene IDQ8N7B1IsotypeIgGELISAYesImmunoChemistryYesImmunogenRecombinant Human HORMA domain-containing protein 2 protein (14-307AA)FormLiquidPurification>95%, Protein G purifiedStorage buffer0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4Storage temperatureUpon receipt, store at –20 or –80 °C. Avoid repeated freeze.Shipping temperatureShipped on blue ice
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



