Anti-GRIK2 Rabbit Polyclonal Antibody
Catalog # 77705-465
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:GRIK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:Q13002
- Antigen synonyms:bA487F5.1|glutamate receptor form D|glutamate receptor form C|GLR 6|GLR6|glutamate receptor form B|EAA4|Glutamate receptor ionotropic kainate 2|GluR6|GLUK6|MRT6|Excitatory amino acid receptor 4|GluR-6|GRIK2_HUMAN|GluK2|GRIK2|glutamate receptor form A|GLUR 6|glutamate receptor form E|Glutamate receptor 6|GRIK2 protein|GRIK 2|ionotropic kainate 2|Glutamate receptor
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthetic Peptide
- Purification:Antigen affinity purified
- Cat. no.:77705-465
- Packaging:Vial
Specifications
About this item
IHC:1:100-200
Type:
Antigen: GRIK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human