Anti-TNR Rabbit Polyclonal Antibody
Catalog # 77705-424
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:TNR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:Q92752
- Antigen synonyms:TN-R|Restrictin|MGC149328|Tnr|Tenascin R (restrictin, janusin)|Janusin|Tenascin-R|TENR_HUMAN
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthetic peptide from Human protein at AA range: 1270-1350
- Purification:Antigen affinity purified
- Cat. no.:77705-424
- Packaging:Vial
Specifications
About this item
IHC:1:50-300, ELISA:1:10000-20000
Type:
Antigen: TNR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat