Anti-OR51I1 Rabbit Polyclonal Antibody
77705-411
New Product
- Antigen symbol:OR51I1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q9H343
- Antigen synonyms:OR51I1|Olfactory receptor 51I1|Odorant receptor HOR5'beta11|Olfactory receptor OR11-39
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the C-terminal region of Human Olfactory receptor 51I1
- Purification:Antigen affinity purified
- Cat. no.:77705-411
- Packaging:Vial
WB:1:500-1:2000, IF:1:200-1:1000, ELISA:1:10000
Type:
Antigen: OR51I1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human