Anti-COL28A1 Rabbit Polyclonal Antibody
Catalog # 77705-389
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:COL28A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q2UY09
- Antigen synonyms:Collagen alpha-1(XXVIII) chain|COL28|Collagen|Col28a1|COSA1_HUMAN|type XXVIII alpha 1
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Collagen alpha-1(XXVIII)Polyclonal Antibody at AA range: 111-160
- Purification:Antigen affinity purified
- Cat. no.:77705-389
- Packaging:Vial
Specifications
About this item
WB:1:500-2000, ELISA:1:10000-20000
Type:
Antigen: COL28A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse