Anti-MMP7 Rabbit Polyclonal Antibody
Catalog # 77715-691
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:MMP7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P09237
- Antigen synonyms:Matrix metalloproteinase-7|Uterine matrilysin|MMP7_HUMAN|MMP7|Pump 1 protease|Uterine metalloproteinase|Matrilysin|PUMP1|Pump-1 protease|Matrin|Matrix Metalloproteinase 7|PUMP 1|MPSL1|MMP-7|MMP 7
- Storage buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Human MMP-7
- Purification:Antigen affinity purified
- Cat. no.:77715-691
- Packaging:Vial
Specifications
About this item
WB:1:500-1:3000, IHC:1:50-1:100
Type:
Antigen: MMP7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat