Anti-TCFL5 Rabbit Polyclonal Antibody
Catalog # 77714-968
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:TCFL5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9UL49
- Antigen synonyms:TCFL5_HUMAN|Cha transcription factor|bHLHe82|Figlb|HPV-16 E2-binding protein 1|transcription factor-like 5 (basic helix-loop-helix)|TCFL 5|E2BP-1|TCFL5|CHA|E2BP1|MGC46135|Transcription factor-like 5 protein
- Storage buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from internal of Human TCFL5
- Purification:Antigen affinity purified
- Cat. no.:77714-968
- Packaging:Vial
Specifications
About this item
WB:1:500-1:3000
Type:
Antigen: TCFL5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse