Anti-LAMA1 Rabbit Polyclonal Antibody
77714-744
New Product
- Antigen symbol:LAMA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µl
- Form:Liquid
- Gene ID:P25391
- Antigen synonyms:Laminin-3 subunit alpha|Laminin subunit alpha-1|LAMA|Laminin A chain|LAMA1|Laminin-1 subunit alpha|S-laminin subunit alpha|S-LAM alpha
- Storage buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from internal of Human LAMA1
- Purification:Antigen affinity purified
- Cat. no.:77714-744
- Packaging:Vial
IF:1:100-1:500
Type:
Antigen: LAMA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human