Anti-STEAP3 Rabbit Polyclonal Antibody
77714-727
New Product
- Antigen symbol:STEAP3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q658P3
- Antigen synonyms:Metalloreductase STEAP3|TSAP 6|Dudulin 2|Dudulin2|STEA 3|STEAP family member 3 metalloreductase|TSAP6|STEAP family member 3|hpHyde|Six transmembrane epithelial antigen of prostate 3|Tumor suppressor-activated pathway protein 6|Six-transmembrane epithelial antigen of prostate 3|hTSAP6|Dudlin 2|STEAP 3|STEA3_HUMAN|Tumor suppressor activated pathway protein 6|pHyde|STEAP3|1010001D01Rik|Dudulin-2|Dudlin2|Tumor suppressor pHyde|STMP 3|STMP3|MGC93147|Six transmembrane prostate protein 3
- Storage buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from internal of Human STEA3
- Purification:Antigen affinity purified
- Cat. no.:77714-727
- Packaging:Vial
WB:1:500-1:3000, IF:1:100-1:500
Type:
Antigen: STEAP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat