Anti-CHST13 Rabbit Polyclonal Antibody
77714-161
New Product
- Antigen symbol:CHST13
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q8NET6
- Antigen synonyms:Carbohydrate (chondroitin 4) sulfotransferase 13|C4ST 3|Chondroitin 4 sulfotransferase 3|Chondroitin 4-sulfotransferase 3|CHSTD_HUMAN|Chondroitin 4-O-sulfotransferase 3|C4ST-3|MGC119278|CHST13|CHST 13|MGC119279|C4ST3|Chondroitin 4 O sulfotransferase 3|MGC119281|Carbohydrate sulfotransferase 13
- Storage buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from internal of Human CHST13
- Purification:Antigen affinity purified
- Cat. no.:77714-161
- Packaging:Vial
WB:1:500-1:3000, IHC:1:50-1:100
Type:
Antigen: CHST13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human