Anti-LRP10 Rabbit Polyclonal Antibody
Catalog # 77713-918
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:LRP10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:100 µl
- Form:Liquid
- Gene ID:Q7Z4F1
- Antigen synonyms:UNQ389/PRO724Low-density lipoprotein receptor-related protein 10|LRP-10|SP220|MSTP087|LRP10
- Storage buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from internal of Human LRP10
- Purification:Antigen affinity purified
- Cat. no.:77713-918
- Packaging:Vial
Specifications
About this item
IHC:1:50-1:100
Type:
Antigen: LRP10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse