Anti-RB1 (phospho-Ser807) Rabbit Polyclonal Antibody
77712-748
New Product
- Antigen symbol:RB1 (phospho-Ser807)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P06400
- Antigen synonyms:Osteosarcoma|RB1|RB_HUMAN|pRb|Retinoblastoma 1|PP105|OSRC|Protein phosphatase 1 regulatory subunit 130|Prepro retinoblastoma associated protein|RB1 gene|Retinoblastoma suspectibility protein|P105RB|GOS563 exon 17 substitution mutation causes premature stop|Rb|Exon 17 tumor GOS561 substitution mutation causes premature stop|pp110|PPP1R130|RB transcriptional corepressor 1|Retinoblastoma-associated protein|p105-Rb
- Storage buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Peptide sequence around phosphorylation site of serine 807 (Y-I-S(p)-P-L) derived from Human Rb
- Purification:Antigen affinity purified
- Cat. no.:77712-748
- Packaging:Vial
WB:1:500-1:1000, IHC:1:50-1:100
Type:
Antigen: RB1 (phospho-Ser807)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat