Anti-ANP32C Rabbit Polyclonal Antibody
77712-143
New Product
- Antigen symbol:ANP32C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:O43423
- Antigen synonyms:pp32 related 1|ANP32 C|Tumorigenic protein pp32r1|Phosphoprotein 32-related protein 1|Phosphoprotein 32 related protein 1|PP32R1|Acidic (leucine rich) nuclear phosphoprotein 32 family, member C|ANP32C|Acidic leucine-rich nuclear phosphoprotein 32 family member C|AN32C_HUMAN
- Storage buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from internal of Human AN32C
- Purification:Antigen affinity purified
- Cat. no.:77712-143
- Packaging:Vial
WB:1:500-1:3000
Type:
Antigen: ANP32C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human