Anti-CSF1R (phospho-Tyr699) Rabbit Polyclonal Antibody
Catalog # 77710-433
Supplier: AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen symbol:CSF1R (phospho-Tyr699)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µl
- Environmentally Preferable:
- Form:Liquid
- Gene ID:P07333
- Antigen synonyms:MCSFR|Oncogen FMS|CD115|CSF-1-R|CSF 1 R|CSF-1 receptor|CSF1 R|CD115 antigen|FIM 2|FMS protooncogene|CSFR|EC 2.7.10.1|CFMS|HDLS|CSF 1R|V-FMS McDonough feline sarcoma viral oncogen homolog|CSF1R_HUMAN|C FMS|CD 115|FMS proto oncogene|Macrophage colony stimulating factor I receptor|Colony stimulating factor 1 receptor|Colony stimulating factor I receptor|M-CSF Receptor|Proto-oncogene c-Fms|Macrophage colony-stimulating factor 1 receptor|McDonough feline sarcoma viral (v fms) oncogene homolog|M-CSF-R|FMS|Macrophage colony stimulating factor 1 receptor|FIM2|CSF1R|formerly
- Storage buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Peptide sequence around phosphorylation site of tyrosine 699 (V-D-Y(p)-K-N) derived from Human CSF1R
- Purification:Antigen affinity purified
- Cat. no.:77710-433
- Packaging:Vial
Specifications
About this item
WB:1:500-1:3000
Type:
Antigen: CSF1R (phospho-Tyr699)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human