Anti-USP42 Rabbit Polyclonal Antibody
77710-362
New Product
- Antigen symbol:USP42
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9H9J4
- Antigen synonyms:UBP42_HUMAN|ubiquitin specific peptidase 42|FLJ12697|Deubiquitinating enzyme 42|Ubiquitin-specific-processing protease 42|Ubiquitin carboxyl-terminal hydrolase 42|UBP42|Ubiquitin thiolesterase 42|USP42
- Storage buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from internal of Human USP42
- Purification:Antigen affinity purified
- Cat. no.:77710-362
- Packaging:Vial
WB:1:500-1:3000
Type:
Antigen: USP42
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse