Anti-SENP1 Rabbit Polyclonal Antibody
Catalog # 77700-544
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:SENP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q9P0U3
- Antigen synonyms:SuPr 2|SuPr2|SENP1|SUMO1/sentrin specific peptidase 1|Sentrin-specific protease 1|SUMO1/sentrin specific protease 1|SENP1_HUMAN|Sentrin/SUMO specific protease SENP1|SENP 1|Sentrin/SUMO specific protease SENP 1|Sentrin/SUMO specific protease|Sentrin/SUMO-specific protease SENP1|Sentrin/SUMO specific protease 1|Sentrin specific protease 1
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the N-terminal region of Human SENP1
- Purification:Antigen affinity purified
- Cat. no.:77700-544
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, IHC:1:100-1:300, ELISA:1:20000
Type:
Antigen: SENP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse