Anti-MOK Rabbit Polyclonal Antibody
Catalog # 77700-451
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:MOK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q9UQ07
- Antigen synonyms:RAGE 1|Antigen recognized by autologous cytolytic T lymphocytes|Renal tumor antigen|MOK_HUMAN|Renal tumor antigen 1|MAPK/MAK/MRK overlapping kinase|MOK protein kinase|EC 2.7.11.22|STK30|LE 9211 A antigen|RAGE-1|Renal cell carcinoma antigen (MOK protein kinase)|RAGE|RAGE1|MOK
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human RAGE
- Purification:Antigen affinity purified
- Cat. no.:77700-451
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, IHC:1:100-1:300, IF:1:200-1:1000, ELISA:1:5000
Type:
Antigen: MOK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human