Anti-PROKR1 Rabbit Polyclonal Antibody
77700-364
New Product
- Antigen symbol:PROKR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q8TCW9
- Antigen synonyms:PK-R1|PROKR1|PKR1|GPR73a|Prokineticin receptor 1|PKR1_HUMAN|zaq|GPR73|PK R1|G protein coupled receptor 73|G-protein coupled receptor ZAQ|EG-VEGF receptor|G-protein coupled receptor 73|G protein coupled receptor ZAQ
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the N-terminal region of Human PKR1
- Purification:Antigen affinity purified
- Cat. no.:77700-364
- Packaging:Vial
WB:1:500-1:2000, IF:1:200-1:1000, ELISA:1:20000
Type:
Antigen: PROKR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat