Anti-PITPNB Rabbit Polyclonal Antibody
Catalog # 77700-347
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:PITPNB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P48739
- Antigen synonyms:PtdIns transfer protein beta|PIPNB_HUMAN|PI TP beta|VIB1B|PI-TP-beta|OTTHUMP00000198846|PtdInsTP beta|Phosphatidylinositol transfer protein beta isoform|Phosphatidylinositol transfer protein, beta|PtdInsTP|Pitpnb|OTTHUMP00000198847
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human PITPβ
- Purification:Antigen affinity purified
- Cat. no.:77700-347
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, ELISA:1:40000
Type:
Antigen: PITPNB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat