Anti-TP53 Rabbit Polyclonal Antibody
77700-264
New Product
- Antigen symbol:TP53
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Monkey,Human
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P04637
- Antigen synonyms:P53_HUMAN|Tp53|Tumor protein 53|Tumor suppressor p53|BCC7|Cellular tumor antigen p53|TRP53|tumor antigen p55|Tumor protein p53|Mutant tumor protein 53|LFS1|Antigen NY-CO-13|Phosphoprotein p53|Transformation related protein 53|FLJ92943|p53|p53 tumor suppressor
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Human p53 around the non-phosphorylation site of S6
- Purification:Antigen affinity purified
- Cat. no.:77700-264
- Packaging:Vial
WB:1:500-1:2000, IHC:1:100-1:300, IF:1:200-1:1000, ELISA:1:20000
Type:
Antigen: TP53
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Monkey