Anti-ACRBP Rabbit Polyclonal Antibody
Catalog # 77700-234
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:ACRBP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q8NEB7
- Antigen synonyms:Cancer/testis antigen OY TES 1|Cancer/testis antigen OY-TES-1|Proacrosin binding protein sp32|FLJ51160|CT23|Proacrosin-binding protein sp32|ACRBP|Acrosin-binding protein|Cancer/testis antigen 23|OY TES 1|ACRBP_HUMAN|Acrosin binding protein precursor|acrosin binding protein|SP32
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human OY-TES-1
- Purification:Antigen affinity purified
- Cat. no.:77700-234
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, IHC:1:100-1:300, IF:1:200-1:1000, ELISA:1:10000
Type:
Antigen: ACRBP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human