Order Entry
Canada
ContactUsLinkComponent
Anti-OR4A16 Rabbit Polyclonal Antibody
Anti-OR4A16 Rabbit Polyclonal Antibody
Catalog # 77700-184
Supplier:  AFG Bioscience
New Product
undefined
Anti-OR4A16 Rabbit Polyclonal Antibody
Catalog # 77700-184
Supplier:  AFG Bioscience
Supplier Number:  A14904-100UG
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antigen symbol:
    OR4A16
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    100 µg
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    Q8NH70
  • Antigen synonyms:
    OR4A16|Olfactory receptor 4A16|Olfactory receptor OR11-117
  • Storage buffer:
    PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
  • Storage temperature:
    Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    Synthesized peptide derived from the C-terminal region of Human Olfactory receptor 4A16
  • Purification:
    Antigen affinity purified
  • Cat. no.:
    77700-184
  • Packaging:
    Vial

Specifications

About this item

WB:1:500-1:2000, IF:1:200-1:1000, ELISA:1:20000

Type:
Antigen: OR4A16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human