Anti-OR1D5/OR1D4 Rabbit Polyclonal Antibody
77700-170
New Product
- Antigen symbol:OR1D5/OR1D4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P58170/P47884
- Antigen synonyms:OR1D5|OR17-31|Olfactory receptor 1D5|OR1D4|Olfactory receptor 1D4|Olfactory receptor 17-30|Olfactory receptor 17-31|OR17-30
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human Olfactory receptor 1D4/1D5
- Purification:Antigen affinity purified
- Cat. no.:77700-170
- Packaging:Vial
WB:1:500-1:2000, IF:1:200-1:1000, ELISA:1:20000
Type:
Antigen: OR1D5/OR1D4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human