Anti-NQO1 Rabbit Polyclonal Antibody
77700-131
New Product
- Antigen symbol:NQO1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P15559
- Antigen synonyms:Diaphorase 4|NMORI|NAD(P)H dehydrogenase quinone 1|Menadione reductase|QR 1|NMOR1|Diaphorase (NADH/NADPH) (cytochrome b 5 reductase)|Dioxin inducible 1|Diaphorase (NADH/NADPH)|NMOR 1|NQO1_HUMAN|NQO1|NMOR I|NAD(P)H:quinone oxireductase|Azoreductase|DIA4|NAD(P)H:quinone oxidoreductase 1|DT diaphorase|NAD(P)H dehydrogenase [quinone] 1|NAD(P)H menadione oxidoreductase 1 dioxin inducible|NAD(P)H quinone dehydrogenase 1|DTD|NAD(P)H:menadione oxidoreductase 1|Phylloquinone reductase|NQO 1|Quinone reductase 1|DHQU|DIA 4|QR1|NAD(P)H:Quinone acceptor oxidoreductase type 1|DT-diaphorase|NAD(P)H: menadione oxidoreductase 1 dioxin inducible 1|Cytochrome b 5 reductase
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the C-terminal region of Human NQO1
- Purification:Antigen affinity purified
- Cat. no.:77700-131
- Packaging:Vial
WB:1:500-1:2000, ELISA:1:10000
Type:
Antigen: NQO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human