Anti-NFKB1 Rabbit Polyclonal Antibody
Catalog # 77700-098
Supplier: AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen symbol:NFKB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P19838
- Antigen synonyms:NFKB1_HUMAN|NFkappaB|DKFZp686C01211|EBP 1|EBP-1|Nfkb1|p50|Nuclear factor NF kappa B p105 subunit|NF kappabeta|Nuclear factor of kappa light chain gene enhancer in B cells 1|NFKB p105|NFKB p50|NFKB-p105|Nuclear factor kappa B DNA binding subunit|Nuclear factor of kappa light polypeptide gene enhancer in B cells 1|DNA binding factor KBF1 EBP1|Nuclear factor NF-kappa-B p50 subunit|nf b|Nuclear factor kappa-B, subunit 1|MGC54151|NF-kappaB|NF kappaB|p84/NF-kappa-B1 p98|DNA-binding factor KBF1|NF kB1|Nuclear factor of kappa light polypeptide gene enhancer in B-cells 1|NFKB 1|NF kappa B|EBP1|p105|KBF1|Transcription factor NFKB1|Nuclear factor kappa-B|DNA binding factor KBF1|Nuclear factor NF kappa B p50 subunit
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Human NFκB-p105/p50 around the non-phosphorylation site of S337
- Purification:Antigen affinity purified
- Cat. no.:77700-098
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, IHC:1:100-1:300, IF:1:200-1:1000, ELISA:1:40000
Type:
Antigen: NFKB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse