Anti-NRP1 Rabbit Polyclonal Antibody
Catalog # 77700-085
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:NRP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:O14786
- Antigen synonyms:NP1|NRP1|Neuropilin-1|VEGF165R|Neuropilin1|NRP1_HUMAN|BDCA4|NPN1|BLOOD DENDRITIC CELL ANTIGEN 4|NRP|transmembrane receptor|Vascular endothelial cell growth factor 165 receptor|A5 protein|NRP 1|CD304
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human Neuropilin-1
- Purification:Antigen affinity purified
- Cat. no.:77700-085
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, IHC:1:100-1:300, ELISA:1:5000
Type:
Antigen: NRP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat