Anti-NBPF12/NBPF10/NBPF1/NBPF9/NBPF20/NBPF15/NBPF14 Rabbit Polyclonal Antibody
Catalog # 77700-052
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:NBPF12/NBPF10/NBPF1/NBPF9/NBPF20/NBPF15/NBPF14
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q5TAG4/Q6P3W6/Q5SXJ2/Q3BBV0/Q3BBW0/Q3BBV1/Q8N660/Q5TI25
- Antigen synonyms:COAS1|Neuroblastoma breakpoint family member 10|Neuroblastoma brea|NBPF1|NBPF10|NBPF12|NBPF16|Neuroblastoma breakpoint family member 16|KIAA1245|KIAA1693|Chromosome 1 amplified sequence 1|Neuroblastoma breakpoint family member 12
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the C-terminal region of Human NBPF1/9/10/12/14/15/16/20
- Purification:Antigen affinity purified
- Cat. no.:77700-052
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, IHC:1:100-1:300, ELISA:1:40000
Type:
Antigen: NBPF12/NBPF10/NBPF1/NBPF9/NBPF20/NBPF15/NBPF14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human