Anti-MAP2K1/MAP2K2 Rabbit Polyclonal Antibody
Catalog # 77699-927
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:MAP2K1/MAP2K2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:Q02750/P36507
- Antigen synonyms:PRKMK2|MEK1|Dual specificity mitogen-activated protein k|MAPKK 1|Dual specificity mitogen-activated protein kinase kinase 1|MAP2K1|MEK 1|ERK activator kinase 1|MEK2|PRKMK1|MKK2|MAP2K2|MAP kinase kinase 1|MAPK/ERK kinase 1|MKK1
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Human MEK-1/2 around the non-phosphorylation site of S218/222
- Purification:Antigen affinity purified
- Cat. no.:77699-927
- Packaging:Vial
Specifications
About this item
Type:
Antigen: MAP2K1/MAP2K2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Monkey