Anti-HOXA7 Rabbit Polyclonal Antibody
Catalog # 77699-722
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:HOXA7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P31268
- Antigen synonyms:Homeobox protein Hox-A7|OTTHUMP00000024835|HOX1.1, mouse, homolog of|HOXA7|Hox1A|Homeobox protein Hox-1A|HOX1|Homeobox 1A|Homeobox protein Hox 1.1|HOX1.1|Homeobox protein Hox A7|Antp, Drosophila, homolog of|Homeobox A7|HXA7_HUMAN|ANTP
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human Hox-A7
- Purification:Antigen affinity purified
- Cat. no.:77699-722
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, ELISA:1:40000
Type:
Antigen: HOXA7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat