Anti-HIST1H2BC/HIST1H2BJ/HIST2H2BE/H2BFS Rabbit Polyclonal Antibody
Catalog # 77699-694
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:HIST1H2BC/HIST1H2BJ/HIST2H2BE/H2BFS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P62807/P06899/Q16778/P57053
- Antigen synonyms:H2B/g|HIST1H2BC|H2BFK|H2B/h|Histone H2B.a|HIST1H2BG|HIST1H2BI|HIST1H2BE|Histone H2B.h|Histone H2B.k|Histone H2B type 1-C/E/F/G/I|Histone H2B.l|Histone H2B.1 A|H2BFH|Histone H2B.g|HIST1H2BF|H2B/k|H2BFA|H2BFG|H2BFL|H2B/a
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Human Histone H2B around the non-acetylation site of K5
- Purification:Antigen affinity purified
- Cat. no.:77699-694
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, IHC:1:100-1:300, IF:1:200-1:1000, ELISA:1:20000
Type:
Antigen: HIST1H2BC/HIST1H2BJ/HIST2H2BE/H2BFS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse