ContactUsLinkComponent
Tablet for Pipetting Applications, TRACKMAN® Connected, Gilson
Tablet for Pipetting Applications, TRACKMAN® Connected, Gilson
Catalog # 76333-068
Supplier:  GILSON, INC.
undefined
Tablet for Pipetting Applications, TRACKMAN® Connected, Gilson
Catalog # 76333-068
Supplier:  GILSON, INC.
Supplier Number:  FB1022
Specifications
  • Description:
    TRACKMAN® Connected tablet, US-plug
  • Cat. no.:
    76333-068
Specifications
About this item
TRACKMAN® Connected is a tablet with exclusive accessories and applications that make pipetting on 96- and 384-well plates faster and more reliable, improving your efficiency at the bench by tracking your pipetting tasks.
  • Adds traceability
  • Boosts experiment reliability
  • Provides immediate reproducibility
Designed to communicate with PIPETMAN® M Connected via Bluetooth®, the tablet interacts in real-time with the pipette and guides you through your protocol with PipettePilot®. When complete, results can be saved, shared, and exported to SciNote, an electronic lab notebook. Easily capture notes, photos, and monitor your protocols. Streamline your work with the Bluetooth-connected environment sensor by automatically registering a protocol run action such as pipetting and conditions like temperature, humidity, and air pressure. When paired with PIPETMAN M Connected, the installed PipettePilot app guides you through your microplate method. The app displays where, when, and how much to pipette with real-time interactions to prevent errors. Effortlessly create and share your protocols. Compatibility with SciNote lets you structure your work into projects, experiments, and tasks, making it easy to visualize your work progress. Sync your results for a comprehensive view of your pipetting actions, samples, and protocols.