ContactUsLinkComponent
Electrode Current USC
Electrode Current USC
Catalog # 77664-240
New Product
Electrode Current USC
Catalog # 77664-240
Supplier Number:  EKC
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Electrode current USC, red tip, 2 mm pin
  • Cat. no.:
    77664-240
Specifications
About this item
Current (red) and voltage (black) electrodes are threaded and screw securely into the end of each Luer tipped cartridge. The Luer tip then plugs securely into the Luer openings of the chamber. The cable from each electrode terminates with a 2 mm pin which may be plugged into most voltage/current clamps.
  • Ussing chamber current electrode
  • Works with most voltage/current clamps
The miniature electrode gel cartridge is a small plastic tube with a male Luer tip identical to those at the tip of hypodermic syringes. The tube may be filled with different gel materials. Agar is commonly used, but other gel materials may also be satisfactory.