If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Description:Powder precision cleaner
- Cat. no.:77661-644
- pH:9.5
- Weight:4 lbs.
Alconox® is a concentrated, anionic detergent for manual and ultrasonic cleaning. This detergent gives you reliable results without interfering residues, because it rinses away easily. Alconox cleaner is Ideal for cleaning contaminants from glassware, metals, plastics, ceramic, porcelain, rubber and fiberglass. It is an excellent replacement for corrosive acids and hazardous solvents.
- Save money with a concentrated powder that can be diluted 1:100
- Protect the environment with this biodegradable and readily disposable cleaning solution
- Save time with penetrating wetting power
- Get reliable results with no interfering residues on labware
- Free rinsing means it rinses away easily for residue-free results
- Corrosion inhibited means Alconox is even safe on soft metals, plastics, rubber and fiberglass
- Use with manual or ultrasonic cleaning methods
- Pass your cleaning validation tests for lab accreditation and plant inspection approval using USDA authorized Alconox
Alconox® can be used as a paste, as a soak or ultrasonically. It can be used on glass, metal, stainless steel, porcelain, ceramic, plastic, rubber and fiberglass. It can also be used on soft metals such as copper aluminum, zinc and magnesium, if they are rinsed promptly.
It can be used to remove: soil, grit, grime, buffing compound, slime, grease, blood, tissue, oils, salts, deposits, particulates, solvents, chemicals, radioisotopes, radioactive contaminants, silicone oils and mold release agents.