Order Entry
Canada
ContactUsLinkComponent
Mini Glue Gun with Glue Sticks
Mini Glue Gun with Glue Sticks
  77661-636
New Product
Mini Glue Gun with Glue Sticks
  77661-636
 :  13316
New Product

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Description:
    Mini glue gun with glue sticks
  • Cat. no.:
    77661-636
  • Weight:
    0.5 kg

 

 

Mini glue gun bonds wood, glass, metals, and many plastics.

  • Easy to use for bonding and large gap filling
  • 110 V, 60 Hz only
  • Curing time: As soon as it cools

Includes: Three sticks of special formula hot melt glue.

 : UL approved.