GrantBio Combi-spin Centrifuge/Vortex Mixer, Grant Instruments
Catalog #: 80076-360
Supplier: GRANT USA INC.
Specifications
- Capacity:12 x 1.5/2.0 mL, 12 x 0.5 mL plus 12 x 0.2 mL
- Description:Combi-spin Centrifuge/Vortex Mixer
- Electrical:115V, 50/60Hz, 0.63A
- Shipping weight:2.7 kg (6 lbs.)
- Max. RCF:700 g
- Dimensions:11.5H x 22.5W x 18D cm (41/2 x 87/8 x 71/8")
- Operating temperature:4 to 45°C
- Speed:2400 rpm
- Time setting:Continuous up to 60 min. or short spin
- Cat. no.:80076-360
- Orbit:3.6 mm (0.14")
- Supplier No.:PCV-2400 USA
Specifications
About this item
Cost-effective, fixed-speed, combined micro centrifuge/vortex mixer for combined or independent centrifuge and mixing applications of microtubes and 0.2ml microtube strips in low volume applications.
- Centrifugation speed: fixed at 2800rpm/50Hz and 3500rpm/60Hz
- Continuous operation or short spin
- Choice of interchangeable rotors for different microtube sizes/combinations and for 0.2ml strips
- Can be used at temperatures from +4ºC to +40ºC
Tubes are loaded into the rotor for simultaneous spinning, then removed for individual mixing in the vortex cup located at the top of the central shaft. In spin-mix-spin applications, the Combi-spin™ can be used with very low reagent volumes representing an overall saving in time, labour and material.
Certifications: CE marked.
Ordering information: Includes three-year manufacturer's warranty. Each unit comes complete with two interchangeable rotors: one holds twelve 1.5/2.0mL microtubes, the other holds twelve 0.5mL plus twelve 0.2mL microtubes. Additional rotors are available separately.
Customers Who Bought This Also Bought