Pathology Workstations, Mortech®
About this item
All stainless steel workstations are equipped with powerful variable speed exhaust systems with full-width exhaust grills near the work area.
- Workstation base is constructed of heavy-gauge Type 304 stainless steel with a No. 4 satin finish
- Adjustable leveling feet are provided for all four legs
The countertop workstation features a 12.7L×30.5W×7.6H cm (5×12×3") integrated sink installed in the upper shelf area. The standard workstation features a 30.5W×35.6D×20.3H cm (12×14×8") sink installed in the recessed work area to allow installation of a waste disposal.
Four two-speed fans draw formaldehyde fumes through filter cartridges that break them down into CO₂ and water. An easily accessible control panel controls ventilation. Units may also be connected to a building exhaust system. Lighting is provided by a top-mounted fluorescent light fixture and two magnifying lamps with 3X magnifiers and incandescent bulbs mounted on flexible arms. Workstations are two-tiered, featuring a 25.4 cm (10") deep upper shelf area with a 38.1 cm (15") deep recessed work area below.
All workstations are equipped with a formalin container with spigot, a stainless steel C-fold paper towel holder, a magnetic instrument holder, and a polyethylene dissecting board.
Ordering information: Motors and filters are not provided for units intended to connect to building exhaust systems. Bases are custom fabricated to customer specifications. Optional accessories such as camera mounts, gas/air valves, and integrated dictation equipment are available. Contact your Avantor sales representative for details.
0 Options Available Below
Product Details & Documents
All stainless steel workstations are equipped with powerful variable speed exhaust systems with full-width exhaust grills near the work area.
- Workstation base is constructed of heavy-gauge Type 304 stainless steel with a No. 4 satin finish
- Adjustable leveling feet are provided for all four legs
The countertop workstation features a 12.7L×30.5W×7.6H cm (5×12×3") integrated sink installed in the upper shelf area. The standard workstation features a 30.5W×35.6D×20.3H cm (12×14×8") sink installed in the recessed work area to allow installation of a waste disposal.
Four two-speed fans draw formaldehyde fumes through filter cartridges that break them down into CO₂ and water. An easily accessible control panel controls ventilation. Units may also be connected to a building exhaust system. Lighting is provided by a top-mounted fluorescent light fixture and two magnifying lamps with 3X magnifiers and incandescent bulbs mounted on flexible arms. Workstations are two-tiered, featuring a 25.4 cm (10") deep upper shelf area with a 38.1 cm (15") deep recessed work area below.
All workstations are equipped with a formalin container with spigot, a stainless steel C-fold paper towel holder, a magnetic instrument holder, and a polyethylene dissecting board.
Ordering information: Motors and filters are not provided for units intended to connect to building exhaust systems. Bases are custom fabricated to customer specifications. Optional accessories such as camera mounts, gas/air valves, and integrated dictation equipment are available. Contact your Avantor sales representative for details.