ContactUsLinkComponent
8293 P100 Particulate Respirator, 3M™
8293 P100 Particulate Respirator, 3M™
  CA56221-028
 :  3M
undefined
8293 P100 Particulate Respirator, 3M™
  CA56221-028
 :  3M
 :  7000002055
  • Description:
    8293 P100 Particulate Respirator
  • Cat. no.:
    CA56221-028
  • Supplier No.:
    70070757474
This respirator is well suited for those who want NIOSH's highest rated filtration efficiency in a maintenance-free respirator. For workplace applications involving OSHA substance-specific particle exposures including lead, cadmium, and arsenic.
  • For workplace applications involving OSHA substance-specific particle exposures including lead, cadmium, and arsenic
  • 3M Cool Flow® exhalation valve, a comfortable faceseal, and adjustable straps
  • Provides minimum filtration efficiency of 99.97% against certain solid and liquid aerosols including oils
Respirator features the 3M Cool Flow® exhalation valve, a comfortable faceseal, and adjustable straps. Provides minimum filtration efficiency of 99.97% against certain solid and liquid aerosols including oils.
Caution: All 3M™ products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product.