SAF-T-FIT™ Plus P1135 Disposable Particulate Respirators, Honeywell Safety
Supplier: Honeywell Safety Products
Certificates
About this item
For extended use with contaminant aerosols containing oil
Ideal for sanding, grinding, sweeping, brazing, torch cutting, soldering, certain welding applications, and hot, dusty, or dry operations. Respirators are equipped with an exhalation valve for ease of breathing and exceptional comfort during extended wear. A full sealing flange provides best fit and protection. Respirators with nuisance level organic vapor filtration are ideal for use in foundries, agriculture applications, petrochemical processes, laboratory applications, and in the food and beverage industry. Latex-free.
Frequently Bought Together