ContactUsLinkComponent
2310 Disposable N99 Premium Particulate Respirator, Moldex®
2310 Disposable N99 Premium Particulate Respirator, Moldex®
Catalog # 56219-030
Supplier:  Moldex/Metric
Small Business Enterprise
   Safer Human and Environmental Health
undefined
2310 Disposable N99 Premium Particulate Respirator, Moldex®
Catalog # 56219-030
Supplier:  Moldex/Metric
Supplier Number:  2310
Small Business Enterprise
   Safer Human and Environmental Health
Specifications
  • Description:
    2310 N99 Premium particulate respirator
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Cat. no.:
    56219-030
  • Supplier No.:
    2310N99
Specifications
About this item
Provides a filter efficiency level of 99% or greater against particulate aerosols free of oil up to 10 times Permissible Exposure Limit (PEL).
  • Exhale valve lets workers breathe easily and stay cool
  • Contoured facepiece features a soft-spun linerSoft foam nose seal fits easily and comfortably without a metal noseband
  • The polymesh shell resists collapsing, even in heat and humidity and keeps the filter clean-looking longer
  • Straps are securely attached so they do not break at the seal
Suitable for most welding fumes. Fits easily under welding hoods. Flame retardants added to the Dura-Mesh® shell.
Caution: User is responsible for checking current OSHA regulations and applicable guidelines.