2500 Disposable N95 Particulate Respirators, Moldex®
56219-028
Small Business Enterprise
- Description:2500 N95 Particulate Respirator
- Environmentally Preferable:Safer Human and Environmental Health
- Cat. no.:56219-028
- Supplier No.:2500N95
Used when oil aerosols are not present in industries such as aluminum reduction, paper mills, coal-fired utility plants, petrochemicals, chemical processing and glass etching
- Naturally contoured for a comfortable, secure fit
- Seals easily and without a metal noseband
- Durable, crush-resistant polymesh outer shell keeps the filter cleaner-looking longer
- Resists collapsing, even in heat and humidity
- Exhale valve reduces breathing resistance and hot air build up
- Disposable
Provides a filter efficiency level of 95% or greater against particulate aerosols free of oil up to 10 times Permissible Exposure Limit (PEL). For relief from nuisance levels (less than OSHA PEL) of acid gas irritants.