ContactUsLinkComponent
2600 Disposable N95 Particulate Respirators, Moldex®
2600 Disposable N95 Particulate Respirators, Moldex®
Catalog # 46610-306
Supplier:  Moldex/Metric
Small Business Enterprise
   Safer Human and Environmental Health
undefined
2600 Disposable N95 Particulate Respirators, Moldex®
Catalog # 46610-306
Supplier:  Moldex/Metric
Supplier Number:  2600N95
Small Business Enterprise
   Safer Human and Environmental Health
Specifications
  • Size:
    Medium/Large
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Cat. no.:
    46610-306
  • Supplier No.:
    2600N95
Specifications
About this item
The particulate respirators are made with Softspun® lining for added comfort and durability.
  • Stretch cloth strap feels cool and comfortable against the skin
  • Featuring a contoured nose bridge with soft foam nose flange, this respirator allows all-day comfort
  • Dura-Mesh® shell will not collapse in heat and humidity
  • Certified to have a filter efficiency of 95% or greater against particulates free of oil
The respirators have a cloth HandyStrap® with a buckle allowing the mask to hang around the neck when not in use.