8271 P95 Particulate Respirators, 3M™
Catalog # CA56621-934
Supplier: 3M
Specifications
- Description:8271 P95 Respirator
- Cat. no.:CA56621-934
- Supplier No.:70070710838
Specifications
About this item
Respirator provides protection against certain solid and liquid particles, including those containing oil.
- Provides protection against certain solid and liquid particles, including those containing oil
- Conformable "M" nose clip for an improved fit
- Cool Flow® exhalation valve to reduce heat and moisture inside the respirator
- Face seal provides comfort and fit over broad range of face sizes
- If used in environments containing oil aerosols, limit use to 40 hours or 30 days
- Provides minimum filtration efficiency of 95%
Suggested uses include grinding, sanding, sawing, bagging, and machining. Comes with the conformable “M” nose clip for an improved fit and Cool Flow® exhalation valve to reduce heat and moisture inside the respirator. Faceseal provides comfort and fit over a broad range of face sizes. If used in environments containing oil aerosols, limit use to 40 hours or 30 days, whichever is first. Provides minimum filtration efficiency of 95%.
Caution: All 3M™ products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product.