Order Entry
Canada
Orders LinkContactUsLinkComponent
qScript™ One-Step qRT-PCR Kits

qScript™ One-Step qRT-PCR Kits

qScript™ One-Step qRT-PCR Kits
Supplier:  Quantabio
Certificates
qScript™ One-Step qRT-PCR Kits

1 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Synthesis of cDNA and PCR amplification are carried out in the same tube without opening between procedures

  • One-step convenience
  • High efficiency and sensitivity
  • Available with reference dye pre-mixed

Kits are available for all real-time PCR instrument platforms including those requiring normalization with ROX™ reference dye. Kits 101414-170 and -172 are for use with platforms not requiring passive reference dye, including Roche LightCycler® 480, Bio-Rad® iQ™, MyiQ™, iQ™5, Opticon®, Chromo4™, Corbett Rotor-Gene®, Eppendorf® Mastercycler®, and Cepheid® SmartCycler®. Kits with ROX™ reference dye are for use with platforms including AB 7000, 7300, 7700, and 7900 and StepOne™. Kits with Low ROX™ reference dye are for use with platforms including AB 7500 and Stratagene® Mx™.

Systems have been optimized to deliver maximum RT-PCR efficiency, sensitivity, and specificity, enabling unbiased co-amplification of low copy transcripts in the presence of higher copy reference genes. The proprietary reaction buffer has been specifically formulated to maximize activities of both reverse transcriptase and Taq DNA polymerase while minimizing the potential for primer-dimer and other non-specific PCR artifacts. Highly specific amplification is crucial to successful qRT-PCR as non-specific product(s) can compete for amplification of the target sequence and impair PCR efficiency. A key component of this kit is AccuStart™ Taq DNA polymerase which contains monoclonal antibodies that bind to the polymerase and keep it inactive prior to the initial PCR denaturation step. Upon heat activation at 95°C, the antibodies denature irreversibly, releasing fully active, unmodified Taq DNA polymerase.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • CA101414-172
  • CA101414-176
  • CA101414-178
  • CA101414-180
  • Application:
  • RT-qPCR probe
  • RT-qPCR probe
  • RT-qPCR probe
  • RT-qPCR probe
  • One step:
  • Yes
  • Yes
  • Yes
  • Yes

Specifications

Product Family Options

One-Step qRT-PCR Kits, with Low ROX™

Product Information

  • Cycler compatibility
    No. of reactions
    Availability
    Price
  • CA101414-178
    qScript™ One-Step qRT-PCR, low ROX™
    qScript™ One-Step qRT-PCR, low ROX™
    Agilent AriaMx, BioRad CFX/iCycler iQ™/MyiQ™/iQ™5, QIAGEN Rotor-Gene Q, Quantabio Q, Roche LightCycler 480
    50
    Discontinued
    Supplier Number: -
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    CA101414-178
    Application
    RT-qPCR probe
    One step
    Yes
  • CA101414-180
    qScript™ One-Step qRT-PCR, low ROX™
    qScript™ One-Step qRT-PCR, low ROX™
    Agilent AriaMx, BioRad CFX/iCycler iQ™/MyiQ™/iQ™5, QIAGEN Rotor-Gene Q, Quantabio Q, Roche LightCycler 480
    200
    Log in for availability
    Log in for availability
    Each
    Each - 1 278,95 $
    1 278,95 $
    Each
       1 278,95 $
    Minimum order quantity is 1

    Supplier Number: 95059-200
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    CA101414-180
    Application
    RT-qPCR probe
    One step
    Yes
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.