Specifications
- Turning angle - reciprocal mode:0 - 360° (step 30°)
- Description:Multi-Function 3D Rocker-Rotator
- Electrical:12 V
- Max. load:1 kg
- Orbit:0 - 5 mm (step 1°), vibration mode
- Platform dimensions:210 x 210 mm
- Ambient temperature range:4...40 °C
- Operating time:168 hrs. (continuous operation)
- Tilt angle:7°
- Display:2 x 16 character LCD
- No. of cycles:0 - 125 (repetitions)
- Speed:1 - 100 rpm
- Timer:0 - 250 sec. (orbital/reciprocal mode)
0 - 5 sec. (vibration mode) - Rocking angle:0 - 5 (step 1°), vibration mode
- Supplier No.:PS-M3D
Specifications
About this item
The PS-M3D is a fixed angle, variable speed, multifunction 3D rotator providing variable rotation, reciprocation and vibration in a compact unit
Combining different phases in a customised sequence gives the following possible combinations: 3D orbital; 3D reciprocal; 3D vibration; 3D orbital and reciprocal, 3D orbital and vibration; 3D orbital and reciprocal and vibration; 3D reciprocal and vibration. An individual programme may be configured for any different combination of components to ensure optimum mixing conditions.
All actions can be set for continuous or timed operation. Individual phases can be linked together for flexibility. Can be used at 4 to 40°C (80% relative humidity, non-condensing), in an incubator or cold room.
Applications include immunoprecipitation, tissue culture treatments, gel staining, destaining, antibody staining, washes, hybridisations, and blots. The stepper motor produces regulated and reproducible rotating throughout the speed range. The PS-M3D provides all that is needed for mixing different sized particles: 3D orbital (long wave) required by large particles; Reciprocal (long and medium wave) ideal for large to medium particles; Vibration (short wave) suits small particles.
Manufacturers 2 year warranty.